5-Bromo-6-chloropyridin-3-ol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR345245-5G
APOSOR345245-5GEA 143 EUR
APOSOR345245-5G
5-Bromo-6-chloropyridin-3-ol 97%
5-Bromo-6-chloropyridin-3-ol
Formule: C₅H₃BrClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09835248
CAS nummer: 130115-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR