5-Amino-2-chloronicotinic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR345344-250MG OR345344-100MG
APOSOR345344-250MGEA 250 EUR
APOSOR345344-250MG APOSOR345344-100MG
5-Amino-2-chloronicotinic acid
5-Amino-2-chloronicotinic acid
Formule: C₆H₅ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06658302
CAS nummer: 42959-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR