N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR346004-1G OR346004-500MG
APOSOR346004-1GEA 0 EUR
APOSOR346004-1G APOSOR346004-500MG
N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formule: C₁₃H₁₈ClN₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10568180
CAS nummer: 159016-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR