3-Oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester 2-ethyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR346102-1G OR346102-5G
APOSOR346102-1GEA 0 EUR
APOSOR346102-1G APOSOR346102-5G
3-Oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester 2-ethyl ester
3-Oxo-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester 2-ethyl ester
Formule: C₁₅H₁₇NO₅
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11052441
CAS nummer: 51814-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR