3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR346184-1G OR346184-5G
APOSOR346184-1GEA 168 EUR
APOSOR346184-1G APOSOR346184-5G
3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester
3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester
Formule: C₇H₁₀N₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12131137
CAS nummer: 103626-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR