(±)-Methyl-2,3-dibromopropionate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3479-5G
APOSOR3479-5GEA 22 EUR
APOSOR3479-5G
(±)-Methyl-2,3-dibromopropionate
(±)-Methyl-2,3-dibromopropionate
Formule: C₄H₆Br₂O₂
Kookpunt: 83…86 °C
Dichtheid: 1,944 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00017882
CAS nummer: 1729-67-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR