(3-Formyl-5-methylphenyl)boronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR350181-1G
APOSOR350181-1GEA 58.4 EUR
APOSOR350181-1G
(3-Formyl-5-methylphenyl)boronic acid
(3-Formyl-5-methylphenyl)boronic acid
Formule: C₈H₉BO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08276762
CAS nummer: 870777-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR