Methyl 1-amino-1H-pyrrole-2-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR350273-1G OR350273-250MG
APOSOR350273-1GEA 284 EUR
APOSOR350273-1G APOSOR350273-250MG
Methyl 1-amino-1H-pyrrole-2-carboxylate
Methyl 1-amino-1H-pyrrole-2-carboxylate
Formule: C₆H₈N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11226170
CAS nummer: 122181-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR