(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR350354-5G
APOSOR350354-5GEA 0 EUR
APOSOR350354-5G
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol
Formule: C₈H₁₃NOS
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07357308
CAS nummer: 116539-55-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR