3-Bromo-6-chloropicolinic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3520-1G OR3520-5G OR3520-10G
APOSOR3520-1GEA 22 EUR
APOSOR3520-1G APOSOR3520-5G APOSOR3520-10G
3-Bromo-6-chloropicolinic acid 97%
3-Bromo-6-chloropicolinic acid
Formule: C₆H₃BrClNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09258777
CAS nummer: 929000-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR