Dibenzothiophene-2-boronic acid 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR360153-5G OR360153-1G OR360153-25G
APOSOR360153-5GEA 52.6 EUR
APOSOR360153-5G APOSOR360153-1G APOSOR360153-25G
Dibenzothiophene-2-boronic acid 99%
Dibenzothiophene-2-boronic acid
Formule: C₁₂H₉BO₂S
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD01318982
CAS nummer: 668983-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR