(4-Chlorocarbonylphenyl)boronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR360238-1G OR360238-25G OR360238-5G
APOSOR360238-1GEA 87.6 EUR
APOSOR360238-1G APOSOR360238-25G APOSOR360238-5G
(4-Chlorocarbonylphenyl)boronic acid
(4-Chlorocarbonylphenyl)boronic acid
Formule: C₇H₆BClO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08062416
CAS nummer: 332154-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR