7-Chloro-2-methylquinoline-3-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3612-1G
APOSOR3612-1GEA 312 EUR
APOSOR3612-1G
7-Chloro-2-methylquinoline-3-carboxylic acid
7-Chloro-2-methylquinoline-3-carboxylic acid
Formule: C₁₁H₈ClNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05863549
CAS nummer: 171270-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR