N-(1H-Imidazol-2-ylmethyl)-4-chloroaniline ≥98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR370089-1G OR370089-5G
APOSOR370089-1GEA 178 EUR
APOSOR370089-1G APOSOR370089-5G
N-(1H-Imidazol-2-ylmethyl)-4-chloroaniline ≥98%
N-(1H-Imidazol-2-ylmethyl)-4-chloroaniline
Formule: C₁₀H₁₀ClN₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08668354
CAS nummer: 166096-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR