ω-Bromo-2-cyanoacetophenone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR370138-1G OR370138-250MG
APOSOR370138-1GEA 935 EUR
APOSOR370138-1G APOSOR370138-250MG
ω-Bromo-2-cyanoacetophenone 95%
ω-Bromo-2-cyanoacetophenone
Formule: C₉H₆BrNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03407832
CAS nummer: 683274-86-4
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR