4-Amino-3-hydroxypyridine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3715-1G
APOSOR3715-1GEA 185 EUR
APOSOR3715-1G
4-Amino-3-hydroxypyridine 97%
4-Amino-3-hydroxypyridine
Formule: C₅H₆N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01941262
CAS nummer: 52334-53-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR