Allylboronic acid pinacol ester 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR3740-5G OR3740-25G OR3740-100G
APOSOR3740-5GEA 22 EUR
APOSOR3740-5G APOSOR3740-25G APOSOR3740-100G
Allylboronic acid pinacol ester 97%
Allylboronic acid pinacol ester
Formule: C₉H₁₇BO₂
Kookpunt: 50…53 °C/5mmHg
Dichtheid: 0,896 g/cm³ (25 °C)
Vlampunt: 46 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013347
CAS nummer: 72824-04-5
UN: 1993
ADR: 3,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR