HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR3890-250G OR3890-25G OR3890-1KG
APOSOR3890-250GEA 29.2 EUR
APOSOR3890-250G APOSOR3890-25G APOSOR3890-1KG
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%
HMDS (1,1,1,3,3,3-hexamethyldisilazane)
Formule: (CH₃)₃SiNHSi(CH₃)₃
Kookpunt: 125…127 °C
Smeltpunt: –80 °C
Dichtheid: 0,77 g/cm³ (25 °C)
Vlampunt: 8 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008259
CAS nummer: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3+6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR