4-[Methoxy(methyl)carbamoyl]benzeneboronic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR3967-100G OR3967-25G OR3967-5G
APOSOR3967-100GEA 843 EUR
APOSOR3967-100G APOSOR3967-25G APOSOR3967-5G
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid 97%
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid
Formule: C₉H₁₂BNO₄
Smeltpunt: 118…124 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115671
CAS nummer: 179055-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR