1-(4-Bromo-2-nitrophenyl)ethanone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR400597-1G
APOSOR400597-1GEA 60 EUR
APOSOR400597-1G
1-(4-Bromo-2-nitrophenyl)ethanone 98%
1-(4-Bromo-2-nitrophenyl)ethanone
Formule: C₈H₆BrNO₃
Kookpunt: 345,9 °C
Smeltpunt: 70 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD11977406
CAS nummer: 90004-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR