3-Chloro-2-hydroxybenzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR400649-1G OR400649-25G OR400649-5G
APOSOR400649-1GEA 24.9 EUR
APOSOR400649-1G APOSOR400649-25G APOSOR400649-5G
3-Chloro-2-hydroxybenzonitrile
3-Chloro-2-hydroxybenzonitrile
Formule: C₇H₄ClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09999455
CAS nummer: 13073-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR