3-Chloro-4-iodobenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR400978-1G OR400978-250MG
APOSOR400978-1GEA 141 EUR
APOSOR400978-1G APOSOR400978-250MG
3-Chloro-4-iodobenzyl alcohol 95%
3-Chloro-4-iodobenzyl alcohol
Formule: C₇H₆ClIO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18393355
CAS nummer: 166386-60-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR