3-Iodo-5-nitrobenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR401062-1G OR401062-250MG OR401062-5G
APOSOR401062-1GEA 33.7 EUR
APOSOR401062-1G APOSOR401062-250MG APOSOR401062-5G
3-Iodo-5-nitrobenzyl alcohol 95%
3-Iodo-5-nitrobenzyl alcohol
Formule: C₇H₆INO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18397990
CAS nummer: 219589-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR