4-Acetyl-2-methylbenzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR40110-1G OR40110-5G
APOSOR40110-1GEA 526 EUR
APOSOR40110-1G APOSOR40110-5G
4-Acetyl-2-methylbenzonitrile
4-Acetyl-2-methylbenzonitrile
Formule: C₁₀H₉NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11845792
CAS nummer: 1138444-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR