Triethylsilylacetylene 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR40699-100G
APOSOR40699-100GEA 173 EUR
APOSOR40699-100G
Triethylsilylacetylene 97%
Triethylsilylacetylene
Formule: C₈H₁₆Si
MW: 140.3 g/mol
Dichtheid: 0.783 g/cm³ (25 °C)
Vlampunt: 17 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075373
CAS nummer: 1777-03-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR