1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR40739-1G OR40739-5G OR40739-25G
APOSOR40739-1GEA 26.3 EUR
APOSOR40739-1G APOSOR40739-5G APOSOR40739-25G
1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%
1-Amino-1-cyclopropanecarbonitrile hydrochloride
Formule: C₄H₇ClN₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04114063
CAS nummer: 127946-77-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR