4-Bromo-3-cyclopropylaniline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR40779-1G OR40779-250MG OR40779-5G
APOSOR40779-1GEA 526 EUR
APOSOR40779-1G APOSOR40779-250MG APOSOR40779-5G
4-Bromo-3-cyclopropylaniline 95%
4-Bromo-3-cyclopropylaniline
Formule: C₉H₁₀BrN
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD21100004
CAS nummer: 1353855-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR