N-Benzyl-2-chloroacetamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR4110-1G
APOSOR4110-1GEA 58.4 EUR
APOSOR4110-1G
N-Benzyl-2-chloroacetamide
N-Benzyl-2-chloroacetamide
Formule: C₉H₁₀ClNO
Smeltpunt: 93…96 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00084929
CAS nummer: 2564-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR