1,3-Benzoxazol-2-amine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR42126-1G OR42126-10G OR42126-5G
APOSOR42126-1GEA 22 EUR
APOSOR42126-1G APOSOR42126-10G APOSOR42126-5G
1,3-Benzoxazol-2-amine 98%
1,3-Benzoxazol-2-amine
Formule: C₇H₆N₂O
Smeltpunt: 129 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00182850
CAS nummer: 4570-41-6
EINECS: 224-952-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR