2,6-Dichloro-p-phenylenediamine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR4321-1G OR4321-5G OR4321-10G
APOSOR4321-1GEA 22 EUR
APOSOR4321-1G APOSOR4321-5G APOSOR4321-10G
2,6-Dichloro-p-phenylenediamine 98%
2,6-Dichloro-p-phenylenediamine
Formule: C₆H₆Cl₂N₂
Smeltpunt: 122…127 °C
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00025369
CAS nummer: 609-20-1
EINECS: 210-184-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR