4-Toluenethiol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Mercapto-4-methylbenzene, 4-Methylbenzene-1-thiol, 4-Thiocresol, 4-Methylbenzenethiol, p-Toluenethiol, p-Thiocresol

OR43548-100G OR43548-500G OR43548-25G
APOSOR43548-100GEA 106 EUR
APOSOR43548-100G APOSOR43548-500G APOSOR43548-25G
4-Toluenethiol
4-Toluenethiol
Formule: C₇H₈S
Kookpunt: 195 °C
Smeltpunt: 40…44 °C
Vlampunt: 68 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004851
CAS nummer: 106-45-6
EINECS: 203-399-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR