6-Bromoindazole-1-carboxylic acid tert-butyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR43564-1G OR43564-5G OR43564-250MG
APOSOR43564-1GEA 43.8 EUR
APOSOR43564-1G APOSOR43564-5G APOSOR43564-250MG
6-Bromoindazole-1-carboxylic acid tert-butyl ester
6-Bromoindazole-1-carboxylic acid tert-butyl ester
Formule: C₁₂H₁₃BrN₂O₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06739133
CAS nummer: 877264-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR