4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR4391-250MG OR4391-1G
APOSOR4391-250MGEA 0 EUR
APOSOR4391-250MG APOSOR4391-1G
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride 97%
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride
Formule: C₁₀H₇ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07368530
CAS nummer: 222541-76-6
UN: 3261
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR