imidazo[1,2-a]pyridine-6-carbonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR4430-1G OR4430-250MG OR4430-5G
APOSOR4430-1GEA 27.8 EUR
APOSOR4430-1G APOSOR4430-250MG APOSOR4430-5G
imidazo[1,2-a]pyridine-6-carbonitrile 97%
imidazo[1,2-a]pyridine-6-carbonitrile
Formule: C₈H₅N₃
Smeltpunt: 160…163 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06659603
CAS nummer: 106850-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR