4-Bromo-2,6-dichlorobenzenesulphonyl chloride 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR4495-25G OR4495-5G OR4495-1G
APOSOR4495-25GEA 0 EUR
APOSOR4495-25G APOSOR4495-5G APOSOR4495-1G
4-Bromo-2,6-dichlorobenzenesulphonyl chloride 96%
4-Bromo-2,6-dichlorobenzenesulphonyl chloride
Formule: C₆H₂BrCl₃O₂S
Smeltpunt: 46…50 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094651
CAS nummer: 351003-54-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR