2-Phenylacetyl isocyanate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR450129-1G OR450129-250MG
APOSOR450129-1GEA 1650 EUR
APOSOR450129-1G APOSOR450129-250MG
2-Phenylacetyl isocyanate
2-Phenylacetyl isocyanate

May form small amount of sediment

Formule: C₉H₇NO₂
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD16301042
CAS nummer: 4461-27-2
UN: 2206
ADR: 6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR