6-Chloro-m-anisidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR4502-5G OR4502-1G
APOSOR4502-5GEA 29.2 EUR
APOSOR4502-5G APOSOR4502-1G
6-Chloro-m-anisidine
6-Chloro-m-anisidine
Formule: C₇H₈ClNO
Kookpunt: 135…137 °C/12mmHg
Smeltpunt: 27 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00047830
CAS nummer: 2401-24-3
EINECS: 219-277-6
UN: 2233
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR