3-(Chloromethyl)-5-[(morpholin-4-yl)methyl]isoxazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR45060-1G OR45060-250MG
APOSOR45060-1GEA 261 EUR
APOSOR45060-1G APOSOR45060-250MG
3-(Chloromethyl)-5-[(morpholin-4-yl)methyl]isoxazole
3-(Chloromethyl)-5-[(morpholin-4-yl)methyl]isoxazole
Formule: C₉H₁₃ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11841078
CAS nummer: 953408-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR