Sodium-1-naphthol-4-sulphonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR45108-1G
APOSOR45108-1GEA 29.5 EUR
APOSOR45108-1G
Sodium-1-naphthol-4-sulphonate
Sodium-1-naphthol-4-sulphonate
Formule: C₁₀H₈O₄S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00021518
CAS nummer: 6099-57-6
EINECS: 228-050-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR