L-Cyclopropylglycine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR451084-5G
APOSOR451084-5GEA 125 EUR
APOSOR451084-5G
L-Cyclopropylglycine
L-Cyclopropylglycine
Formule: C₅H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06659116
CAS nummer: 49606-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR