1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR451160-1G OR451160-5G OR451160-10G
APOSOR451160-1GEA 24.9 EUR
APOSOR451160-1G APOSOR451160-5G APOSOR451160-10G
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 95%
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid
Formule: C₉H₁₄O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11847571
CAS nummer: 66500-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR