(R)-2-Cyclopentylglycine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: H-D-Cyclopentyl-Gly-OH, (R)-2-Amino-2-cyclopentylacetic acid

OR451206-5G
APOSOR451206-5GEA 144 EUR
APOSOR451206-5G
(R)-2-Cyclopentylglycine
(R)-2-Cyclopentylglycine
Formule: C₇H₁₃NO₂
MW: 143.184 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03788075
CAS nummer: 2521-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR