2-Bromo-4-iodo-6-methylphenol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR45128-1G OR45128-250MG OR45128-5G
APOSOR45128-1GEA 228 EUR
APOSOR45128-1G APOSOR45128-250MG APOSOR45128-5G
2-Bromo-4-iodo-6-methylphenol
2-Bromo-4-iodo-6-methylphenol
Formule: C₇H₆BrIO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD27664804
CAS nummer: 1820641-98-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR