α-Furfuryl octanoate 98+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR451301-100G OR451301-25G OR451301-500G
APOSOR451301-100GEA 631 EUR
APOSOR451301-100G APOSOR451301-25G APOSOR451301-500G
α-Furfuryl octanoate 98+%
α-Furfuryl octanoate
Formule: C₁₃H₂₀O₃
MW: 224.296 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00209510
CAS nummer: 39252-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR