Benzyloxyacetaldehyde diethyl acetal 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR451328-10G OR451328-50G OR451328-100G
APOSOR451328-10GEA 48.3 EUR
APOSOR451328-10G APOSOR451328-50G APOSOR451328-100G
Benzyloxyacetaldehyde diethyl acetal 95%
Benzyloxyacetaldehyde diethyl acetal
Formule: C₁₃H₂₀O₃
MW: 224.30 g/mol
Kookpunt: 99…100 °C
Dichtheid: 0.987 g/cm³ (25 °C)
Vlampunt: 79 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051786
CAS nummer: 42783-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR