o-Toluamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Methylbenzamide

OR451329-25G OR451329-100G
APOSOR451329-25GEA 23.4 EUR
APOSOR451329-25G APOSOR451329-100G
o-Toluamide 98%
o-Toluamide
Formule: C₈H₉NO
MW: 135.17
Kookpunt: 254
Smeltpunt: 141…142 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007982
CAS nummer: 527-85-5
Merck Index: 12,09666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR