4-Chloro-D-tryptophan 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR452031-1G OR452031-250MG OR452031-500MG
APOSOR452031-1GEA 986 EUR
APOSOR452031-1G APOSOR452031-250MG APOSOR452031-500MG
4-Chloro-D-tryptophan 97%
4-Chloro-D-tryptophan
Formule: C₁₁H₁₁ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10565751
CAS nummer: 27542-41-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR