N,N'-Dicarbobenzoxy-L-cystine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR452094-25G OR452094-100G
APOSOR452094-25GEA 42.4 EUR
APOSOR452094-25G APOSOR452094-100G
N,N'-Dicarbobenzoxy-L-cystine 97%
N,N'-Dicarbobenzoxy-L-cystine
Formule: C₂₂H₂₄N₂O₈S₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00022030
CAS nummer: 6968-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR