N-Boc-(S)-2-(methylamino)butyric acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (S)-2-((tert-Butoxycarbonyl)(methyl)amino)butanoic acid

OR452097-1G OR452097-5G
APOSOR452097-1GEA 203 EUR
APOSOR452097-1G APOSOR452097-5G
N-Boc-(S)-2-(methylamino)butyric acid 97%
N-Boc-(S)-2-(methylamino)butyric acid
Formule: C₁₀H₁₉NO₄
MW: 217.262 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD17215563
CAS nummer: 101759-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR