(2S)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR452188-1G OR452188-250MG OR452188-500MG
APOSOR452188-1GEA 907 EUR
APOSOR452188-1G APOSOR452188-250MG APOSOR452188-500MG
(2S)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride 97%
(2S)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride
Formule: C₅H₁₄N₂·2ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16619103
CAS nummer: 82105-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR